missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT10 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 462.00
Specifications
| Antigen | CNOT10 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CNOT10 Polyclonal antibody specifically detects CNOT10 in Human, Mouse samples. It is validated for Western BlotSpecifications
| CNOT10 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| CCR4-NOT transcription complex subunit 10, CCR4-NOT transcription complex, subunit 10, DKFZp434K115, FLJ12890, FLJ13165 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 465-744 of human CNOT10 (NP_056257.1). LRNALLLLPEEQQDPKQENGAKNSNQLGGNTESSESSETCSSKSHDGDKFIPAPPSSPLRKQELENLKCSILACSAYVALALGDNLMALNHADKLLQQPKLSGSLKFLGHLYAAEALISLDRISDAITHLNPENVTDVSLGISSNEQDQGSDKGENEAMESSGKRAPQCYPSSVNSARTVMLFNLGSAYCLRSEYDKARKCLHQAASMIHPKEVPPEAILLAVYLELQNGNTQLALQIIKRNQLLPAVKTHSEVRKKPVFQPVHPIQPIQMPAFTTVQRK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 25904 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title