missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99135-100ul
This item is not returnable.
View return policy
Description
CNOT6 Polyclonal antibody specifically detects CNOT6 in Human samples. It is validated for Western Blot
Specifications
| CNOT6 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| carbon catabolite repression 4 protein, Carbon catabolite repressor protein 4 homolog, CCR4 carbon catabolite repression 4-like, CCR4CCR4-NOT transcription complex subunit 6, CCR4-NOT transcription complex, subunit 6, Cytoplasmic deadenylase, EC 3.1.13.4, KIAA1194EC 3.1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human CNOT6. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 57472 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction