missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 307.65
Specifications
| Antigen | CNOT6 |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18659825
|
Novus Biologicals
NBP2-99135-100ul |
100 μg |
€ 326.00 € 307.65 / 100µL Save € 18.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18629995
|
Novus Biologicals
NBP2-99135-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CNOT6 Polyclonal antibody specifically detects CNOT6 in Human samples. It is validated for Western BlotSpecifications
| CNOT6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| carbon catabolite repression 4 protein, Carbon catabolite repressor protein 4 homolog, CCR4 carbon catabolite repression 4-like, CCR4CCR4-NOT transcription complex subunit 6, CCR4-NOT transcription complex, subunit 6, Cytoplasmic deadenylase, EC 3.1.13.4, KIAA1194EC 3.1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human CNOT6. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 57472 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title