missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNPase Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP3-05551-50ul
This item is not returnable.
View return policy
Description
CNPase Polyclonal antibody specifically detects CNPase in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| CNPase | |
| Polyclonal | |
| Unconjugated | |
| 50% PBS, 50% glycerol | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
| Western Blot, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:5000, Immunocytochemistry/ Immunofluorescence 1:3000 | |
| 2', 3' cyclic nucleotide 3' phosphohydrolase, 2'-3'-cyclic nucleotide 3' phosphodiesterase, 2'-3'-cyclic-nucleotide 3'-phosphodiesterase, CNP1, CNPase, EC 3.1.4.37 | |
| Full length human recombinant CNPase expressed in E. coli | |
| 50 μg | |
| Cellular Markers, Glia Markers, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers | |
| 1267 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction