missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNPase Antibody, Novus Biologicals™
Goat Polyclonal Antibody
€ 325.50 - € 528.15
Specifications
| Antigen | CNPase |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:5000, Immunocytochemistry/ Immunofluorescence 1:3000 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18614628
|
Novus Biologicals
NBP3-05551-100ul |
100 μg |
€ 559.00 € 528.15 / 100µL Save € 30.85 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18655996
|
Novus Biologicals
NBP3-05551-50ul |
50 μg |
€ 345.00 € 325.50 / 50µL Save € 19.50 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CNPase Polyclonal antibody specifically detects CNPase in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| CNPase | |
| Western Blot 1:5000, Immunocytochemistry/ Immunofluorescence 1:3000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine | |
| 2', 3' cyclic nucleotide 3' phosphohydrolase, 2'-3'-cyclic nucleotide 3' phosphodiesterase, 2'-3'-cyclic-nucleotide 3'-phosphodiesterase, CNP1, CNPase, EC 3.1.4.37 | |
| Full length human recombinant CNPase expressed in E. coli | |
| Immunogen affinity purified |
| 1 mg/mL | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Goat | |
| Cellular Markers, Glia Markers, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers | |
| 50% PBS, 50% glycerol | |
| 1267 | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title