missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor XI Antibody (MM0193-7C38) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12199
This item is not returnable.
View return policy
Description
Coagulation Factor XI Monoclonal specifically detects Coagulation Factor XI in Human samples. It is validated for Western Blot, Block/Neutralize.
Specifications
| Coagulation Factor XI | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000, Block/Neutralize | |
| P03951 | |
| F11 | |
| Human recombinant Coagulation Factor XI | |
| 0.1 mg | |
| Primary | |
| Detects human Coagulation factor XI (Light Chain) | |
| Human | |
| Purified |
| Western Blot, Blocking Assay | |
| MM0193-7C38 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
| coagulation factor XI, EC 3.4.21, EC 3.4.21.27, FXIPlasma thromboplastin antecedent, MGC141891, PTA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 2160 | |
| Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction