missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor XI Antibody (MM0193-7C38) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 195.00 - € 520.00
Specifications
| Antigen | Coagulation Factor XI |
|---|---|
| Clone | MM0193-7C38 |
| Concentration | LYOPH |
| Applications | Western Blot, Blocking Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18028936
|
Novus Biologicals
NBP2-12199-0.025MG |
0.025 mg |
€ 195.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18006016
|
Novus Biologicals
NBP2-12199 |
0.1 mg |
€ 520.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Coagulation Factor XI Monoclonal specifically detects Coagulation Factor XI in Human samples. It is validated for Western Blot, Block/Neutralize.Specifications
| Coagulation Factor XI | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| P03951 | |
| 2160 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0193-7C38 | |
| Western Blot, Blocking Assay | |
| Unconjugated | |
| Mouse | |
| Human | |
| coagulation factor XI, EC 3.4.21, EC 3.4.21.27, FXIPlasma thromboplastin antecedent, MGC141891, PTA | |
| F11 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title