Learn More
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 2mm Thick
Shop All Fisherbrand Products
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
12
14
16
18
2 Prep, 2 Marker
20
28
30
35
4 Prep, 2 Marker
Volume (Metric) Sample:
20 μL
23 μL
25 μL
43 μL
54 μL
59 μL
81 μL
90 μL
243 μL
450 μL
990 μL
Unit Size:
Each
Description
- Recommended for high resolution gels and publication quality data
- Maximise sample volume
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible
Specifications
Specifications
| Color | Blue |
| For Use With (Equipment) | MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit |
| Type | Comb |
| No. of Wells | 1 Prep, 1 Marker |
| Quantity | 1 Each |
| Thickness (Metric) | 2 mm |
| Volume (Metric) Sample | 990 μL |