Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 0.75mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
- Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
- Recommended for ultra resolved bands
- 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
Black | |
OmniPAGE Mini Vertical Gel Electrophoresis Unit | |
8 | |
0.75 mm |
Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations | |
Comb | |
1 Each | |
40 μL |