missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 0.75mm Thick

For use in preparatory electrophoresis and high-throughput screening.

Brand:  Fisherbrand™

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11897593

  • € 18.85 / Each
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

  • Accessory for use with OmniPAGE mini vertical gel electrophoresis unit
  • Recommended for ultra resolved bands
  • 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications

Specifications

Black
OmniPAGE Mini Vertical Gel Electrophoresis Unit
16
0.75 mm
Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations
Comb
1 Each
20 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers