Learn More
COMMD5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10393-1-AP-150UL
 View more versions of this product
Product Code. 16856753
  • € 410.00 / 150µL
Description
May modulate activity of cullin-RING E3 ubiquitin ligase (CRL) complexes. Negatively regulates cell proliferation. Negatively regulates cell cycle G2/M phase transition probably by transactivating p21/CDKN1A through the p53/TP53-independent signaling pathway. Involved in kidney proximal tubule morphogenesis. Down-regulates activation of NF-kappa-B.
Specifications
Specifications
COMMD5
Polyclonal
Unconjugated
COMMD5
COMM domain containing 5, COMMD5, HCARG, HT002
Rabbit
Antigen Affinity Chromatography
RUO
28991, 66398
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.34 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8R395, Q9GZQ3
COMMD5
COMMD5 Fusion Protein Ag0592
150 μL
Primary
Human, Mouse