missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COMMD5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | COMMD5 |
|---|---|
| Concentration | 0.34 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16866753
|
Proteintech
10393-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16856753
|
Proteintech
10393-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May modulate activity of cullin-RING E3 ubiquitin ligase (CRL) complexes. Negatively regulates cell proliferation. Negatively regulates cell cycle G2/M phase transition probably by transactivating p21/CDKN1A through the p53/TP53-independent signaling pathway. Involved in kidney proximal tubule morphogenesis. Down-regulates activation of NF-kappa-B.Specifications
| COMMD5 | |
| Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q8R395, Q9GZQ3 | |
| 28991, 66398 | |
| COMMD5 Fusion Protein Ag0592 | |
| Primary | |
| -20°C | |
| COMMD5 |
| 0.34 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| COMM domain containing 5, COMMD5, HCARG, HT002 | |
| COMMD5 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title