Learn More
WTW™ Connection Cable for Depth Probes
For use with WTW™ MPP IDS depth probes with waterproof plug head.
Brand: WTW™ 903854
Additional Details : Weight : 0.02000kg
Description
- Also useful for plug head sensors
- Available in a range of lengths
- Supplied individually
Specifications
Connection Cable | |
15 m | |
MPP IDS with waterproof head sensors |