missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 43/GJA1 Rabbit anti-Human, Clone: 105, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89209-100UL
This item is not returnable.
View return policy
Description
Connexin 43/GJA1 Monoclonal specifically detects Connexin 43/GJA1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Connexin 43/GJA1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 105 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500 | |
| connexin 43, connexin-43, Cx43, CX43gap junction protein, alpha-like, DFNB38, Gap junction 43 kDa heart protein, gap junction alpha-1 protein, gap junction protein, alpha 1, 43kDa, gap junction protein, alpha 1, 43kDa (connexin 43), GJAL, HSS, ODD, ODDD, ODOD, SDTY3 | |
| A synthetic peptide corresponding to the center region of the Human Connexin 43/GJA1 | |
| 100 μg | |
| Cancer, Cellular Markers, Core ESC Like Genes, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 2697 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction