missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 43/GJA1 Rabbit anti-Human, Clone: 105, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 392.00 - € 578.00
Specifications
| Antigen | Connexin 43/GJA1 |
|---|---|
| Clone | 105 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18215097
|
Novus Biologicals
NBP2-89209-50UL |
50 μg |
€ 392.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18223577
|
Novus Biologicals
NBP2-89209-100UL |
100 μg |
€ 578.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Connexin 43/GJA1 Monoclonal specifically detects Connexin 43/GJA1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Connexin 43/GJA1 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| connexin 43, connexin-43, Cx43, CX43gap junction protein, alpha-like, DFNB38, Gap junction 43 kDa heart protein, gap junction alpha-1 protein, gap junction protein, alpha 1, 43kDa, gap junction protein, alpha 1, 43kDa (connexin 43), GJAL, HSS, ODD, ODDD, ODOD, SDTY3 | |
| A synthetic peptide corresponding to the center region of the Human Connexin 43/GJA1 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 105 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, Core ESC Like Genes, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 0.2 um filtered solution in PBS | |
| 2697 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title