missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin-4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92013-0.1ml
This item is not returnable.
View return policy
Description
Contactin-4 Polyclonal antibody specifically detects Contactin-4 in Human samples. It is validated for Western Blot
Specifications
| Contactin-4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| AXCAM, axonal-associated cell adhesion molecule, BIG-2Brain-derived immunoglobulin superfamily protein 2, contactin 4, contactin-4, MGC33615, neural cell adhesion protein BIG-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 448-697 of human CNTN4 (NP_001193885.1). VGVFNNKGEGPFSPTTVVYSAEEEPTKPPASIFARSLSATDIEVFWASPLEKNRGRIQGYEVKYWRHEDKEENARKIRTVGNQTSTKITNLKGSVLYHLAVKAYNSAGTGPSSATVNVTTRKPPPSQPPGNIIWNSSDSKIILNWDQVKALDNESEVKGYKVLYRWNRQSSTSVIETNKTSVELSLPFDEDYIIEIKPFSDGGDGSSSEQIRIPKISNAYARGSGASTSNACTLSAISTIMISLTARSSL | |
| 0.1 mL | |
| Cell Biology, Neuroscience | |
| 152330 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction