missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin-4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 463.00
Specifications
| Antigen | Contactin-4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18670741
|
Novus Biologicals
NBP2-92013-0.02ml |
0.02 mL |
€ 190.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18626560
|
Novus Biologicals
NBP2-92013-0.1ml |
0.1 mL |
€ 463.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Contactin-4 Polyclonal antibody specifically detects Contactin-4 in Human samples. It is validated for Western BlotSpecifications
| Contactin-4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 152330 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| AXCAM, axonal-associated cell adhesion molecule, BIG-2Brain-derived immunoglobulin superfamily protein 2, contactin 4, contactin-4, MGC33615, neural cell adhesion protein BIG-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 448-697 of human CNTN4 (NP_001193885.1). VGVFNNKGEGPFSPTTVVYSAEEEPTKPPASIFARSLSATDIEVFWASPLEKNRGRIQGYEVKYWRHEDKEENARKIRTVGNQTSTKITNLKGSVLYHLAVKAYNSAGTGPSSATVNVTTRKPPPSQPPGNIIWNSSDSKIILNWDQVKALDNESEVKGYKVLYRWNRQSSTSVIETNKTSVELSLPFDEDYIIEIKPFSDGGDGSSSEQIRIPKISNAYARGSGASTSNACTLSAISTIMISLTARSSL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title