missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin-4 Antibody (SR1190), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22476-50ul
This item is not returnable.
View return policy
Description
Contactin-4 Monoclonal antibody specifically detects Contactin-4 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Contactin-4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1190 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| AXCAM, axonal-associated cell adhesion molecule, BIG-2Brain-derived immunoglobulin superfamily protein 2, contactin 4, contactin-4, MGC33615, neural cell adhesion protein BIG-2 | |
| A synthesized peptide derived from human Contactin-4 (Uniprot #: Q8IWV2) | |
| 50 μg | |
| Cell Biology, Neuroscience | |
| 152330 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction