missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contactin-4 Antibody (SR1190), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 472.00
Specifications
| Antigen | Contactin-4 |
|---|---|
| Clone | SR1190 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625684
|
Novus Biologicals
NBP3-22476-100ul |
100 μg |
€ 472.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649863
|
Novus Biologicals
NBP3-22476-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Contactin-4 Monoclonal antibody specifically detects Contactin-4 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| Contactin-4 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AXCAM, axonal-associated cell adhesion molecule, BIG-2Brain-derived immunoglobulin superfamily protein 2, contactin 4, contactin-4, MGC33615, neural cell adhesion protein BIG-2 | |
| A synthesized peptide derived from human Contactin-4 (Uniprot #: Q8IWV2) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1190 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Neuroscience | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 152330 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title