missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Nickel Aluminum Bronze, CDA 955, C95500, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Nickel Aluminum Bronze, CDA 955, C95500, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Carbon/Boron/Antimony/Arsenic/Calcium/Sulfur/Oxygen/Bismuth/Cadmium/Hydrogen/Silver/Cobalt/Tin/Lead/Chromium/Phosphorus/Silicon/Zinc/Manganese/Iron/Nickel/Aluminium/Copper/Palladium/Nitrogen/Selenium/Titanium/Zirconium/Magnesium/Molybdenum/Vanadium |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Carbon -80 ppm/Boron -30 ppm/Antimony -20 ppm/Arsenic -20 ppm/Calcium -20 ppm/Sulfur -12 ppm/Oxygen -10 ppm/Bismuth -5 ppm/Cadmium -3 ppm/Hydrogen -1 ppm/Silver 0.0019 %/Cobalt 0.0027 %/Tin 0.0056 %/Lead 0.006 %/Chromium 0.008 %/Phosphorus 0.013 %/Silicon 0.022 %/Zinc 0.038 %/Manganese 0.77 %/Iron 3.5 %/Nickel 4.5 %/Aluminium 10.37 %/Copper 80.8 %/Palladium (<1) ppm/Nitrogen (<10) ppm/Selenium (<10) ppm/Titanium (<10) ppm/Zirconium (<10) ppm/Magnesium (<5) ppm/Molybdenum (<5) ppm/Vanadium (<50) ppm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction