missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Naval Brass, Medium Leaded, CDA 482, C48200, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, Naval Brass, Medium Leaded, CDA 482, C48200, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Cobalt/Chromium/Bismuth/Cadmium/Manganese/Phosphorus/Arsenic/Silicon/Antimony/Nickel/Iron/Lead/Tin/Titanium/Copper/Beryllium/Niobium/Zirconium/Boron/Magnesium/Nitrogen/Oxygen/Carbon/Aluminium/Tellurium/Silver/Selenium |
| CAS | 7440-50-8 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Cobalt 0.0007 %/Chromium 0.0009 %/Bismuth 0.0012 %/Cadmium 0.0015 %/Manganese 0.0024 %/Phosphorus 0.004 %/Arsenic 0.005 %/Silicon 0.005 %/Antimony 0.007 %/Nickel 0.013 %/Iron 0.06 %/Lead 0.42 %/Tin 0.69 %/Titanium 38.1 %/Copper 60.7 %/Beryllium (<0.001) %/Niobium (<0.001) %/Zirconium (<0.001) %/Boron (<0.1) %/Magnesium (0.0003) %/Nitrogen (0.0006) %/Oxygen (0.0007) %/Carbon (0.002) %/Aluminium (0.003) %/Tellurium (0.003) %/Silver (0.004) %/Selenium (0.005) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction