missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Aluminum Bronze, CDA 623, C62300, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Aluminum Bronze, CDA 623, C62300, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Arsenic/Carbon/Phosphorus/Tin/Zinc/Silicon/Manganese/Nickel/Iron/Aluminium/Copper/Oxygen/Chromium/Nitrogen/Antimony/Cobalt/Lead/Silver |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Arsenic 0.003 %/Carbon 0.003 %/Phosphorus 0.006 %/Tin 0.01 %/Zinc 0.014 %/Silicon 0.033 %/Manganese 0.2 %/Nickel 0.55 %/Iron 2.28 %/Aluminium 9.2 %/Copper 87.6 %/Oxygen (0.001) %/Chromium (0.002) %/Nitrogen <0.0001 %/Antimony <0.005 %/Cobalt <0.005 %/Lead <0.005 %/Silver <0.005 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction