missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Aluminum Bronze, CDA 955M, C95510, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Copper, Aluminum Bronze, CDA 955M, C95510, 100g
Specifications
Specifications
| Chemical Name or Material | Silver/Carbon/Phosphorus/Lead/Tin/Silicon/Zinc/Manganese/Iron/Nickel/Aluminium/Copper/Nitrogen/Oxygen/Beryllium/Bismuth/Cadmium/Magnesium/Niobium/Boron/Arsenic/Cobalt/Antimony/Chromium/Selenium |
| CAS | 7440-50-8 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Silver 0.0013 %/Carbon 0.005 %/Phosphorus 0.005 %/Lead 0.006 %/Tin 0.019 %/Silicon 0.075 %/Zinc 0.122 %/Manganese 0.6 %/Iron 3.7 %/Nickel 4.7 %/Aluminium 9.91 %/Copper 80.8 %/Nitrogen (0.0004) %/Oxygen (0.0006) %/Beryllium (0.001) %/Bismuth (0.001) %/Cadmium (0.001) %/Magnesium (0.001) %/Niobium (0.001) %/Boron (0.002) %/Arsenic (0.003) %/Cobalt (0.003) %/Antimony (0.004) %/Chromium (0.004) %/Selenium (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction