missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Brass Copper Zinc Binary, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, Brass Copper Zinc Binary, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Boron/Cobalt/Arsenic/Cadmium/Manganese/Silicon/Antimony/Nickel/Lead/Aluminium/Bismuth/Tin/Iron/Zinc/Copper/Tellurium |
| CAS | 7440-50-8 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0013 %/Cobalt 0.0044 %/Arsenic 0.0054 %/Cadmium 0.0064 %/Manganese 0.0088 %/Silicon 0.018 %/Antimony 0.0196 %/Nickel 0.0351 %/Lead 0.0416 %/Aluminium 0.0435 %/Bismuth 0.0607 %/Tin 0.089 %/Iron 0.099 %/Zinc 15.34 %/Copper 84.22 %/Tellurium (0.002) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction