missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Brass Majors, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Brass Majors, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Arsenic/Antimony/Bismuth/Silicon/Tin/Lead/Aluminium/Iron/Nickel/Manganese/Zinc/Copper |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Arsenic 0.0181 %/Antimony 0.0194 %/Bismuth 0.0198 %/Silicon 0.0207 %/Tin 0.0229 %/Lead 0.0244 %/Aluminium 0.081 %/Iron 0.43 %/Nickel 0.491 %/Manganese 1.72 %/Zinc 36.67 %/Copper 60.51 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction