missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Copper Nickel 30%, CDA 715, C71500, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Copper Nickel 30%, CDA 715, C71500, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Sulfur/Cobalt/Tin/Carbon/Magnesium/Phosphorus/Titanium/Silicon/Iron/Manganese/Nickel/Copper/Hydrogen/Tellurium/Boron/Cadmium/Molybdenum/Niobium/Selenium/Vanadium/Zirconium/Antimony/Lead/Nitrogen/Oxygen/Silver/Chromium/Aluminium/Bismuth/Zinc/Arsenic |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Sulfur 0.002 %/Cobalt 0.004 %/Tin 0.004 %/Carbon 0.005 %/Magnesium 0.005 %/Phosphorus 0.009 %/Titanium 0.073 %/Silicon 0.09 %/Iron 0.51 %/Manganese 0.8 %/Nickel 31 %/Copper 67.4 %/Hydrogen (<0.0010) %/Tellurium (<0.0020) %/Boron (<0.0050) %/Cadmium (<0.0050) %/Molybdenum (<0.0050) %/Niobium (<0.0050) %/Selenium (<0.0050) %/Vanadium (<0.0050) %/Zirconium (<0.0050) %/Antimony (0.001) %/Lead (0.001) %/Nitrogen (0.001) %/Oxygen (0.001) %/Silver (0.001) %/Chromium (0.002) %/Aluminium (0.004) %/Bismuth (0.004) %/Zinc (0.007) %/Arsenic (0.01) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction