missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Nickel Aluminum Bronze, CDA 955, C95500, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
Each
Description
Copper, Nickel Aluminum Bronze, CDA 955, C95500, 100g
Specifications
Specifications
| Chemical Name or Material | Silver/Cobalt/Tin/Lead/Chromium/Phosphorus/Silicon/Zinc/Manganese/Iron/Nickel/Aluminium/Copper/Antimony/Arsenic/Bismuth/Boron/Cadmium/Calcium/Carbon/Hydrogen/Magnesium/Molybdenum/Nitrogen/Oxygen/Palladium/Selenium/Sulfur/Titanium/Vanadium/Zirconium |
| CAS | 7440-50-8 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Silver 0.0019 %/Cobalt 0.0027 %/Tin 0.0056 %/Lead 0.006 %/Chromium 0.008 %/Phosphorus 0.013 %/Silicon 0.022 %/Zinc 0.038 %/Manganese 0.77 %/Iron 3.5 %/Nickel 4.5 %/Aluminium 10.37 %/Copper 80.8 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction