missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Oxygen-Free Electronic Copper, CDA 101, C10100, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Copper, Oxygen-Free Electronic Copper, CDA 101, C10100, 100g
Specifications
Specifications
| Chemical Name or Material | Manganese/Bismuth/Magnesium/Zinc/Lead/Antimony/Arsenic/Selenium/Tin/Nickel/Iron/Sulfur/Silver/Cobalt/Chromium/Cadmium/Aluminium/Nitrogen/Oxygen/Silicon/Phosphorus/Carbon/Tellurium/Copper |
| CAS | 7440-50-8 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Manganese 0.00003 %/Bismuth 0.00005 %/Magnesium 0.00006 %/Zinc 0.00008 %/Lead 0.00012 %/Antimony 0.00014 %/Arsenic 0.00015 %/Selenium 0.0002 %/Tin 0.0002 %/Nickel 0.00027 %/Iron 0.0003 %/Sulfur 0.0005 %/Silver 0.0013 %/Cobalt (0.00003) %/Chromium (0.00005) %/Cadmium (0.00007) %/Aluminium (0.0001) %/Nitrogen (0.0001) %/Oxygen (0.0002) %/Silicon (0.0003) %/Phosphorus (0.0004) %/Carbon (0.001) %/Tellurium (0.001) %/Copper (99.93) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction