missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Phosphor Bronze 5% A, CDA 510, C51000, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Phosphor Bronze 5% A, CDA 510, C51000, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Iron/Nickel/Carbon/Antimony/Zinc/Lead/Phosphorus/Tin/Copper/Nitrogen/Aluminium/Arsenic/Manganese/Oxygen/Silver/Silicon/Bismuth |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Iron 0.002 %/Nickel 0.002 %/Carbon 0.003 %/Antimony 0.005 %/Zinc 0.007 %/Lead 0.016 %/Phosphorus 0.148 %/Tin 4.66 %/Copper 95.2 %/Nitrogen (0.0001) %/Aluminium (0.001) %/Arsenic (0.001) %/Manganese (0.002) %/Oxygen (0.002) %/Silver (0.002) %/Silicon (0.003) %/Bismuth (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction