missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Phosphor Bronze B-2, CDA 544, C54400, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Phosphor Bronze B-2, CDA 544, C54400, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Silver/Antimony/Iron/Nickel/Phosphorus/Zinc/Lead/Tin/Copper/Bismuth/Aluminium/Manganese/Carbon/Silicon/Arsenic/Cobalt |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Silver 0.008 %/Antimony 0.01 %/Iron 0.02 %/Nickel 0.077 %/Phosphorus 0.19 %/Zinc 3.55 %/Lead 3.87 %/Tin 4.73 %/Copper 87.7 %/Bismuth (0.001) %/Aluminium (0.002) %/Manganese (0.002) %/Carbon <0.002 %/Silicon <0.002 %/Arsenic <0.003 %/Cobalt <0.005 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction