missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Red Brass, Gun Metal, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
Each
Description
Copper, Red Brass, Gun Metal, 50g
Specifications
Specifications
| Chemical Name or Material | Cadmium/Tellurium/Bismuth/Cobalt/Silicon/Sulfur/Silver/Arsenic/Phosphorus/Aluminium/Antimony/Iron/Nickel/Tin/Lead/Zinc/Copper |
| CAS | 7440-50-8 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Cadmium 0.0048 %/Tellurium 0.0075 %/Bismuth 0.018 %/Cobalt 0.0298 %/Silicon 0.031 %/Sulfur 0.0411 %/Silver 0.0497 %/Arsenic 0.0498 %/Phosphorus 0.0507 %/Aluminium 0.055 %/Antimony 0.072 %/Iron 0.127 %/Nickel 0.728 %/Tin 4.48 %/Lead 5.18 %/Zinc 5.66 %/Copper 83.39 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction