missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Tin Zinc Bismuth Selenium, EnviroBrass 2-2 (SeBiLOY II*), C89520M, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Copper, Tin Zinc Bismuth Selenium, EnviroBrass 2-2 (SeBiLOY II*), C89520M, 100g
Specifications
Specifications
| Chemical Name or Material | Cobalt/Manganese/Oxygen/Aluminium/Silicon/Arsenic/Carbon/Phosphorus/Silver/Lead/Iron/Nickel/Selenium/Bismuth/Zinc/Tin/Copper/Nitrogen/Chromium/Antimony |
| CAS | 7440-50-8 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Cobalt 0.001 %/Manganese 0.001 %/Oxygen 0.0013 %/Aluminium 0.002 %/Silicon 0.002 %/Arsenic 0.003 %/Carbon 0.003 %/Phosphorus 0.003 %/Silver 0.004 %/Lead 0.042 %/Iron 0.06 %/Nickel 0.53 %/Selenium 1.21 %/Bismuth 2.3 %/Zinc 4.7 %/Tin 5.1 %/Copper 85.9 %/Nitrogen (0.0002) %/Chromium (0.001) %/Antimony <0.01 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction