missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Zinc Bismuth Selenium, EnviroBrass 3 (SeBiLOY III*), C89550M, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Copper, Zinc Bismuth Selenium, EnviroBrass 3 (SeBiLOY III*), C89550M, 50g
Specifications
Specifications
| Chemical Name or Material | Selenium/Cadmium/Arsenic/Phosphorus/Cobalt/Antimony/Lead/Iron/Silicon/Aluminium/Nickel/Bismuth/Tin/Zinc/Copper |
| CAS | 7440-50-8 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Selenium 0.0027 %/Cadmium 0.0055 %/Arsenic 0.0224 %/Phosphorus 0.0232 %/Cobalt 0.0437 %/Antimony 0.087 %/Lead 0.118 %/Iron 0.143 %/Silicon 0.151 %/Aluminium 0.504 %/Nickel 0.55 %/Bismuth 1.031 %/Tin 1.267 %/Zinc 35.01 %/Copper 61.12 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction