missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Zinc Bismuth Selenium, EnviroBrass 3 (SeBiLOY III*), C89550M, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Copper, Zinc Bismuth Selenium, EnviroBrass 3 (SeBiLOY III*), C89550M, 50g
Specifications
Specifications
| Chemical Name or Material | Cadmium/Cobalt/Selenium/Antimony/Arsenic/Iron/Silicon/Phosphorus/Nickel/Aluminium/Lead/Tin/Bismuth/Zinc/Copper |
| CAS | 7440-50-8 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Cadmium 0.0014 %/Cobalt 0.0032 %/Selenium 0.0047 %/Antimony 0.0321 %/Arsenic 0.0476 %/Iron 0.051 %/Silicon 0.0516 %/Phosphorus 0.0626 %/Nickel 0.127 %/Aluminium 0.154 %/Lead 0.181 %/Tin 0.198 %/Bismuth 4.04 %/Zinc 31.83 %/Copper 63.18 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction