missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated IDE Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech CL488-67106-20UL

 View more versions of this product

Product Code. 30237007

  • € 140.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The IDE antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IDE. This antibody recognizes human, mouse, rat antigen. The IDE antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications

Specifications

IDE
Monoclonal
1000 μg/mL
FLJ35968; INSULYSIN
Recombinant Protein
20 μL
Primary
Human, Mouse, Rat
100-120 kDa
Liquid
Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining)
3E10G1
CoraLite Plus 488
Mouse
Protein A purification
RUO
3416
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Conjugated Antibody
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.