missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated IDE Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

€ 140.00 - € 450.00

Specifications

Antigen IDE
Clone 3E10G1
Concentration 1000 μg/mL
Applications Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30235476
View Documents
Proteintech
CL488-67106-100UL
100 μL
€ 450.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30237007
View Documents
Proteintech
CL488-67106-20UL
20 μL
€ 140.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The IDE antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IDE. This antibody recognizes human, mouse, rat antigen. The IDE antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications

Specifications

IDE
1000 μg/mL
Monoclonal
Liquid
RUO
3416
Recombinant Protein
Primary
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
3E10G1
Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining)
CoraLite Plus 488
Mouse
Human, Mouse, Rat
FLJ35968; INSULYSIN
IgG2b
Protein A purification
Conjugated Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.