missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated LDHA-Specific Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

€ 140.00 - € 450.00

Specifications

Antigen LDHA-Specific
Concentration 1000 μg/mL
Applications Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining)
Classification Polyclonal
Conjugate CoraLite Plus 488
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30234223
View Documents
Proteintech
CL488-19987-100UL
100 μL
€ 450.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30237292
View Documents
Proteintech
CL488-19987-20UL
20 μL
€ 140.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The LDHA-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human LDHA-Specific. This antibody recognizes human, mouse, rat antigen. The LDHA-Specific antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications

Specifications

LDHA-Specific
Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining)
CoraLite Plus 488
Rabbit
Human, Mouse, Rat
LDH-M; LDH1; PIG19
IgG
Antigen affinity purification
Conjugated Antibody
1000 μg/mL
Polyclonal
Liquid
RUO
3939
Peptide
Primary
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.