missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated LSM12 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67029-20UL
This item is not returnable.
View return policy
Description
The LSM12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LSM12. This antibody recognizes human, mouse, rat antigen. The LSM12 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| LSM12 | |
| Monoclonal | |
| 1000 μg/mL | |
| FLJ30656; MGC104211; PNAS-135 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 22 kDa | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| 2E3G3 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 124801 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction