missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated LSM12 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 140.00 - € 450.00
Specifica
| Antigen | LSM12 |
|---|---|
| Clone | 2E3G3 |
| Concentration | 1000 μg/mL |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|
30236173
|
Proteintech
CL488-67029-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234664
|
Proteintech
CL488-67029-20UL |
20 μL |
€ 140.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
The LSM12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LSM12. This antibody recognizes human, mouse, rat antigen. The LSM12 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.Specifica
| LSM12 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 124801 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 2E3G3 | |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Mouse | |
| Human, Mouse, Rat | |
| FLJ30656; MGC104211; PNAS-135 | |
| IgG2a | |
| Protein A purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title