missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated SQLE Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67206-20UL
This item is not returnable.
View return policy
Description
CoraLite™ Plus 488-conjugated SQLE Mouse Monoclonal Antibody, Proteintech shows reactivity in Human, Rat, Pig samples.Specifications
| SQLE | |
| Monoclonal | |
| 1000 μg/mL | |
| FLJ30795 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Rat, Pig | |
| 50-64 kDa | |
| Liquid | |
| IgG2a |
| Not Available | |
| 1C9A2 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 6713 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| 25 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction