missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated SQLE Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 133.74 - € 409.50
Specifications
| Antigen | SQLE |
|---|---|
| Clone | 1C9A2 |
| Concentration | 1000 μg/mL |
| Applications | Not Available |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234548
|
Proteintech
CL488-67206-100UL |
100 μL |
€ 409.50
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236928
|
Proteintech
CL488-67206-20UL |
20 μL |
€ 133.74
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CoraLite™ Plus 488-conjugated SQLE Mouse Monoclonal Antibody, Proteintech shows reactivity in Human, Rat, Pig samples.Specifications
| SQLE | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 6713 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1C9A2 | |
| Not Available | |
| CoraLite Plus 488 | |
| Mouse | |
| Human, Rat, Pig | |
| FLJ30795 | |
| IgG2a | |
| Protein A purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title