missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX18 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-30593
This item is not returnable.
View return policy
Description
COX18 Polyclonal specifically detects COX18 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| COX18 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Q8N8Q8 | |
| COX18 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: AYQHYILAKVENLQPEIKTIARHLNQEVAVRANQLGWSKRDARLTYLKNMRRLISELYVRDNC | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| COX18 cytochrome c oxidase assembly homolog (S. cerevisiae), COX18HS, Cox18Hs1 protein, Cox18Hs2 protein, Cox18Hs3 protein, Cytochrome c oxidase assembly protein 18, FLJ38991, MGC126733, mitochondrial COX18, mitochondrial inner membrane protein COX18, OXA1L2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 285521 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction