missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX20 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25752-1-AP-20UL
This item is not returnable.
View return policy
Description
COX20, also name as FAM36A, belongs to the COX20 family. Mutations in COX20 are a novel cause of recessively inherited.Specifications
| COX20 | |
| Polyclonal | |
| Unconjugated | |
| COX20 | |
| FAM36A, Protein FAM36A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116228 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5RI15 | |
| COX20 | |
| COX20 Fusion Protein Ag22520 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction