Learn More
Cole-Parmer™ CPC™ Quick-Disconnect Panel-Mount Valved Hose Barb
Ideal for use with gases or liquids
€ 41.95 - € 77.35
Specifications
Product Type | Quick-Disconnect Panel-Mount Hose Barb Body |
---|---|
Description | Quick-Disconnect x Barb |
Material | Brass |
Min. Operating Temperature (English) | -40°F |
Min. Operating Temperature (Metric) | -40°C |
Product Code | Brand | Tubing Size | Outlet Port | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Outlet Port | Price | Quantity & Availability | ||||
15437213
|
Cole-Parmer™
31403-72 |
1/4 in. | 1/4 in. |
€ 77.35
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
||||
15437243
|
Cole-Parmer™
31404-26 |
1/4 in. | 1/8 in. |
€ 45.75
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
||||
15417243
|
Cole-Parmer™
31404-24 |
1/8 in. | 1/8 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately).
- Withstands vacuum to 250psi.
- Temperature range: -40 to +180°F.
Specifications
Quick-Disconnect Panel-Mount Hose Barb Body | |
Brass | |
-40°C | |
180°F |
Quick-Disconnect x Barb | |
-40°F | |
250 psi | |
82°C |