missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPPED1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145456
This item is not returnable.
View return policy
Description
Protein phosphatase that dephosphorylates AKT family kinase specifically at 'Ser-473', blocking cell cycle progression and promoting cell apoptosis. May play an inhibitory role in glucose uptake by adipocytes.
Specifications
| CPPED1 | |
| Polyclonal | |
| Unconjugated | |
| CPPED1 | |
| C530044N13Rik; calcineurin like phosphoesterase domain containing 1; calcineurin-like phosphoesterase domain containing 1; calcineurin-like phosphoesterase domain-containing protein 1; complete S transactivated protein 1; complete S-transactivated protein 1; Cpped1; CSTP1; RGD1306568; Serine/threonine-protein phosphatase CPPED1; Vcalcineurin-like phosphoesterase domain containing 1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 223978, 302890, 55313 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q66H71, Q8BFS6, Q9BRF8 | |
| CPPED1 | |
| Recombinant Mouse CPPED1 protein, Met1-Glu312 (Accession #Q8BFS6). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction