Learn More
DWK Life Sciences Crimp Cap, ND11
Crimp closures generally known for excellent sealing characteristics, even when it comes to long-term sample storage.
Brand: DWK Life Sciences 225683
8.65 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Additional Details : Weight : 0.03200kg
Description
The quality of a crimp seal is strongly dependent on the right calibration of the crimping tool used to apply the closure to the vial. A too tight or too loose crimp will impair the quality of the seal. To maintain good results, make sure to re-calibrate your crimping tool regularly and check the quality of the seal every time you crimp.
- Packed in zip-lock PE-bags with tear-off foil strip to ensure tamper-evidence and re-closeability.
Specifications
Silver | |
With liner | |
ND20 |
Natural Red Rubber, Clear FEP | |
Open Top Cap |