Learn More
Thermo Scientific Chemicals Crizotinib
Product Code. 16594324 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16594324 1 g 1g
16504334 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16594324 Supplier Thermo Scientific Chemicals Supplier No. 465690010
Request Bulk or Custom Format
CAS: 877399-52-5 | C21H22Cl2FN5O | 450.34 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 877399-52-5
Molecular Formula C21H22Cl2FN5O
Molecular Weight (g/mol) 450.34
MDL Number MFCD12407409
InChI Key KTEIFNKAUNYNJU-UHFFFAOYNA-N
IUPAC Name 3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]-5-[1-(piperidin-4-yl)-1H-pyrazol-4-yl]pyridin-2-amine
SMILES CC(OC1=C(N)N=CC(=C1)C1=CN(N=C1)C1CCNCC1)C1=C(Cl)C=CC(F)=C1Cl
Specifications
Quantity 1 g
Formula Weight 450.34
Chemical Name or Material Crizotinib