Learn More
Invitrogen™ CRMP1 Recombinant Rabbit Monoclonal Antibody (23GB3480), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA552560
 View more versions of this product
Product Code. 30088554
  • € 468.00 / 100µL
Description
Collapsin-response mediator proteins (CRMPs) are highly expressed in the developing brain where they play major roles in axonal outgrowth, neurite differentiation, and apoptosis. Their continued expression in areas of high synaptic remodeling such as the cerebellum, hippocampus, and the olfactory system suggests that these proteins may also be involved in adult brain plasticity. CRMP-1 was initially identified as a dihydro-pyrimidinase expressed exclusively in brain; later studies have shown that it is involved with neurotrophin (NT) 3-induced neurite formation and outgrowth. CRMP-1 localization switches from axonal to somatodendritic when neurons reach functional maturity, suggesting that it is involved in early neuronal differentiation as well as in later processes related to the survival or death of the newly generated neurons.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CRMP1
Recombinant Monoclonal
Unconjugated
CRMP1
Collapsin response mediator protein 1; Crmp1; CRMP-1; dihydropyrimidinase-like 1; dihydropyrimidinase-related protein 1; Dpysl1; DRP1; DRP-1; Ulip3; ULIP-3; Unc-33-like phosphoprotein 3
Rabbit
Affinity chromatography
RUO
1400
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB3480
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q14194
CRMP1
A synthetic peptide of human CRMP1 (500-572AA).