missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRMP1 Recombinant Rabbit Monoclonal Antibody (23GB3480), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 195.00 - € 468.00
Specifications
| Antigen | CRMP1 |
|---|---|
| Clone | 23GB3480 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088554
![]() |
Invitrogen™
MA552560 |
100 μL |
€ 468.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088224
|
Invitrogen™
MA552561 |
20 μL |
€ 195.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CRMP1 Recombinant Monoclonal antibody specifically detects CRMP1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| CRMP1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q14194 | |
| 1400 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3480 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Collapsin response mediator protein 1; Crmp1; CRMP-1; dihydropyrimidinase-like 1; dihydropyrimidinase-related protein 1; Dpysl1; DRP1; DRP-1; Ulip3; ULIP-3; Unc-33-like phosphoprotein 3 | |
| A synthetic peptide of human CRMP1 (500-572AA). | |
| Primary | |
| -20°C | |
| CRMP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title