missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRTAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21432-25ul
This item is not returnable.
View return policy
Description
CRTAP Polyclonal antibody specifically detects CRTAP in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| CRTAP | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| cartilage associated protein, cartilage-associated protein, CASPLEPREL3, leprecan-like 3 | |
| Produced in rabbits immunized with E. coli-derived Human CRTAP fragment | |
| 25 μg | |
| Signal Transduction | |
| 10491 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?